Nitrosiumtetrafluoroborat

Lieferant: FLUOROCHEM

Synonyme: Nitrosyltetrafluoroborat

002855-10G 002855-50G
FLUO002855-10GEA 168 EUR
FLUO002855-10G FLUO002855-50G
Nitrosiumtetrafluoroborat
Nitrosium tetrafluoroborate
Formel: BF₄NO
Molecular Weight: 116,81 g/mol
MDL: MFCD00011433
CAS-Nummer: 14635-75-7
EINECS: 238-679-2
UN: 3260
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR