2,2,2-Trifluorethylmethacrylat
Lieferant: FLUOROCHEM
Synonyme:
Methylpropensäure(2,2,2-trifluorethyl)ester, Methacrylsäure(2,2,2-trifluorethyl)ester
003325-25G
003325-100G
003325-500G
FLUO003325-25GEA
0
EUR
FLUO003325-25G
FLUO003325-100G
FLUO003325-500G
2,2,2-Trifluorethylmethacrylat
2,2,2-Trifluorethylmethacrylat
Formel:
C₆H₇F₃O₂ Molecular Weight: 168.12 g/mol |
MDL:
MFCD00013576 CAS-Nummer: 352-87-4 EINECS: 206-525-3 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...