Phenylsulfonylaceton

Lieferant: FLUOROCHEM

Synonyme: 1-(Phenylsulfonyl)aceton

005323-5G 005323-25G
FLUO005323-5GEA 60.9 EUR
FLUO005323-5G FLUO005323-25G
Phenylsulfonylaceton
Phenylsulfonylaceton
Formel: C₉H₁₀O₃S
Molecular Weight: 198.24 g/mol
MDL: MFCD00047794
CAS-Nummer: 5000-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR