2-Fluorphenylhydraziniumchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-Fluorphenylhydrazin Hydrochlorid, 1-(2-Fluorphenyl)hydrazin Hydrochlorid, 2-Fluorphenylhydrazinhydrochlorid

CFF1147. 006332-5G 006332-25G 006332-100G
CHMPCFF1147.EA 0 EUR
CHMPCFF1147. FLUO006332-5G FLUO006332-25G FLUO006332-100G
2-Fluorphenylhydraziniumchlorid
2-Fluorphenylhydraziniumchlorid
Formel: C₆H₈ClFN₂
Molecular Weight: 162.59 g/mol
MDL: MFCD00012927
CAS-Nummer: 2924-15-4
EINECS: 220-885-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR