2-Hydroxy-3-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

Synonyme: 3-(Trifluormethyl)pyridin-2-ol

CFF649.0.5 CFF649.1 006628-1G 006628-5G
CHMPCFF649.0.5EA 508 EUR
CHMPCFF649.0.5 CHMPCFF649.1 FLUO006628-1G FLUO006628-5G
2-Hydroxy-3-(trifluormethyl)pyridin
2-Hydroxy-3-(trifluoromethyl)pyridine
Formel: C₆H₄F₃NO
Molecular Weight: 163.1 g/mol
MDL: MFCD00153194
CAS-Nummer: 22245-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR