2-(Trifluormethyl)phenylhydrazin
Lieferant: FLUOROCHEM
Synonyme:
1-(2-(Trifluormethyl)phenyl)hydrazin, 2-Hydrazinobenzotrifluorid
007645-1G
007645-5G
FLUO007645-1GEA
121
EUR
FLUO007645-1G
FLUO007645-5G
2-(Trifluormethyl)phenylhydrazin
2-(Trifluormethyl)phenylhydrazin
Formel:
C₇H₇F₃N₂ Molecular Weight: 176,14 g/mol |
MDL:
MFCD00052687 CAS-Nummer: 365-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...