3-(Trifluormethyl)phenylhydraziniumchlorid

Lieferant: FLUOROCHEM

Synonyme: 3-(Trifluormethyl)phenylhydrazin Hydrochlorid, 3-Hydrazinobenzotrifluorid Hydrochlorid, 3-(Trifluormethyl)phenylhydrazinhydrochlorid, 1-(3-(Trifluormethyl)phenyl)hydrazin Hydrochlorid

007646-1G 007646-5G
FLUO007646-1GEA 31.1 EUR
FLUO007646-1G FLUO007646-5G
3-(Trifluormethyl)phenylhydraziniumchlorid
3-(Trifluormethyl)phenylhydraziniumchlorid
Formel: C₇H₈ClF₃N₂
Molecular Weight: 212,6 g/mol
MDL: MFCD00100503
CAS-Nummer: 3107-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR