2-(Trifluormethyl)phenylhydraziniumchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluormethyl)phenylhydrazin Hydrochlorid, 2-(Trifluormethyl)phenylhydrazinhydrochlorid, 2-Hydrazinobenzotrifluorid Hydrochlorid, 1-(2-(Trifluormethyl)phenyl)hydrazin Hydrochlorid

CFF1289. 008441-5G 008441-25G
CHMPCFF1289.EA 0 EUR
CHMPCFF1289. FLUO008441-5G FLUO008441-25G
2-(Trifluormethyl)phenylhydraziniumchlorid
2-(Trifluormethyl)phenylhydraziniumchlorid
Formel: C₇H₈ClF₃N₂
MDL: MFCD00102619
CAS-Nummer: 3107-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR