2-Ethylphenylhydraziniumchlorid

Lieferant: FLUOROCHEM

Synonyme: 1-(2-Ethylphenyl)hydrazin Hydrochlorid, 2-Ethylphenylhydrazin Hydrochlorid

008798-25G 008798-10G 008798-100G
FLUO008798-25GEA 128 EUR
FLUO008798-25G FLUO008798-10G FLUO008798-100G
2-Ethylphenylhydraziniumchlorid
2-Ethylphenylhydraziniumchlorid
Formel: C₈H₁₃ClN₂
Molecular Weight: 172.66 g/mol
MDL: MFCD00071599
CAS-Nummer: 19398-06-2
EINECS: 421-460-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR