Cyclopropyl-4-fluorphenylketon

Lieferant: FLUOROCHEM

Synonyme: Cyclopropyl(4-fluorophenyl)methanone

010277-5G 010277-25G
FLUO010277-5GEA 0 EUR
FLUO010277-5G FLUO010277-25G
Cyclopropyl-4-fluorphenylketon
Cyclopropyl-4-fluorphenylketon
Formel: C₁₀H₉FO
Molecular Weight: 164,18 g/mol
MDL: MFCD00013728
CAS-Nummer: 772-31-6
EINECS: 212-247-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR