4-Chlorphenylsulfonylaceton

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Chlorphenylsulfonyl)aceton

011124-25G 011124-100G
FLUO011124-25GEA 168 EUR
FLUO011124-25G FLUO011124-100G
4-Chlorphenylsulfonylaceton
4-Chlorphenylsulfonylaceton
Formel: C₉H₉ClO₃S
Molecular Weight: 232.69 g/mol
MDL: MFCD00018663
CAS-Nummer: 5000-48-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR