2,5-Difluorphenylhydrazin
Lieferant: FLUOROCHEM
013824-25G
013824-5G
FLUO013824-25GEA
0
EUR
FLUO013824-25G
FLUO013824-5G
2,5-Difluorphenylhydrazin
2,5-Difluorphenylhydrazin
Formel:
C₆H₆F₂N₂ Molecular Weight: 144,12 g/mol |
MDL:
MFCD00013384 CAS-Nummer: 97108-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...