2,4,6-Trifluorphenylhydrazin

Lieferant: CHEMPUR

FL13875.POR 013875-1G
CHMPFL13875.POREA 0 EUR
CHMPFL13875.POR FLUO013875-1G
2,4,6-Trifluorphenylhydrazin
2,4,6-Trifluorphenylhydrazin
Formel: C₆H₅F₃N₂
MDL: MFCD03095329
CAS-Nummer: 80025-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR