3,4-Difluorphenylhydraziniumchlorid
Lieferant: FLUOROCHEM
Synonyme:
3,4-Difluorphenylhydrazin Hydrochlorid, 3,4-Difluorphenylhydrazinhydrochlorid, 1-(3,4-Difluorphenyl)hydrazin Hydrochlorid
013916-5G
013916-25G
FLUO013916-5GEA
36.2
EUR
FLUO013916-5G
FLUO013916-25G
3,4-Difluorphenylhydraziniumchlorid
3,4-Difluorphenylhydraziniumchlorid
Formel:
C₆H₇ClF₂N₂ Molecular Weight: 180.59 g/mol |
MDL:
MFCD03094170 CAS-Nummer: 875664-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...