2-Hydroxy-4-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluormethyl)pyridin-2-ol

CFF1181. 017165-1G 017165-500MG
CHMPCFF1181.EA 0 EUR
CHMPCFF1181. FLUO017165-1G FLUO017165-500MG
2-Hydroxy-4-(trifluormethyl)pyridin
2-Hydroxy-4-(trifluoromethyl)pyridine
Formel: C₆H₄F₃NO
Molecular Weight: 163.1 g/mol
MDL: MFCD00153195
CAS-Nummer: 50650-59-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR