4-Nitrobenzolthiol

Lieferant: FLUOROCHEM

Synonyme: 1-Mercapto-4-nitrobenzol, 4-Nitrobenzolthiol, 4-Mercaptonitrobenzol, p-Nitrothiophenol, 4-Nitrophenylmercaptan, 4-Nitrobenzol-1-thiol

019486-10G 019486-50G
FLUO019486-10GEA 0 EUR
FLUO019486-10G FLUO019486-50G
4-Nitrobenzolthiol
4-Nitrothiophenol
Formel: C₆H₅NO₂S
Molecular Weight: 155.18 g/mol
MDL: MFCD00007343
CAS-Nummer: 1849-36-1
EINECS: 217-436-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR