Methyl-2-amino-4,5-dimethoxybenzoat

Lieferant: FLUOROCHEM

Synonyme: Methyl-4,5-dimethoxyanthranilat, 2-Amino-4,5-dimethoxybenzoesäuremethylester

020200-25G 020200-100G
FLUO020200-25GEA 40.1 EUR
FLUO020200-25G FLUO020200-100G
Methyl-2-amino-4,5-dimethoxybenzoat
Methyl-2-amino-4,5-dimethoxybenzoat
Formel: C₁₀H₁₃NO₄
Molecular Weight: 211.22 g/mol
MDL: MFCD00014904
CAS-Nummer: 26759-46-6
EINECS: 247-967-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR