Methyl-2,3-dimethoxybenzoat
Lieferant: FLUOROCHEM
Synonyme:
2,3-Dimethoxybenzoesäuremethylester
021798-25G
021798-100G
FLUO021798-25GEA
69.8
EUR
FLUO021798-25G
FLUO021798-100G
Methyl-2,3-dimethoxybenzoat
Methyl-2,3-dimethoxybenzoat
Formel:
C₁₀H₁₂O₄ Molecular Weight: 196,2 g/mol |
MDL:
MFCD00027555 CAS-Nummer: 2150-42-7 EINECS: 218-425-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...