4-Nitrophenylhydrazinhydrochlorid ≥95%

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1-(4-Nitrophenyl)hydrazin Hydrochlorid, 4-Nitrophenylhydraziniumchlorid, 4-Nitrophenylhydrazin Hydrochlorid

023917-25G 023917-50G
FLUO023917-25GEA 39.9 EUR
FLUO023917-25G FLUO023917-50G
4-Nitrophenylhydrazinhydrochlorid ≥95%
4-Nitrophenylhydrazinhydrochlorid
Formel: C₆H₇N₃O₂·HCl
Molecular Weight: 189.6 g/mol
Schmelzpunkt: 198 °C (Zers.)
MDL: MFCD00040592
CAS-Nummer: 636-99-7
EINECS: 211-273-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR