1,2-Dihydroxybenzol-3,5-disulfonsäure Dinatriumsalz Monohydrat

Lieferant: MP Biomedicals
Achtung

Synonyme: 4,5-Dihydroxy-1,3-benzoldisulfonsäure Dinatriumsalz Monohydrat, Dinatrium-4,5-dihydroxybenzol-1,3-disulfonat Monohydrat, Brenzcatechin-3,5-disulfonsäure Dinatriumsalz Monohydrat, Tiron Monohydrat

0215774105 0215774180 0215774125
ICNA0215774105EA 37.1 EUR
ICNA0215774105 ICNA0215774180 ICNA0215774125
1,2-Dihydroxybenzol-3,5-disulfonsäure Dinatriumsalz Monohydrat
1,2-Dihydroxybenzol-3,5-disulfonsäure Dinatriumsalz Monohydrat

Colorimetric reagent for iron, manganese, titanium, and molybdenum. Complexing agent.


Very freely soluble in water

Formel: C₆H₄Na₂O₈S₂·1H₂O
Molecular Weight: 332.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149531
CAS-Nummer: 149-45-1
EINECS: 205-741-5

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR