2-Brom-4-fluorbenzonitril

Lieferant: CHEMPUR

FL13212.POR 013212-KIT 013212-25G
CHMPFL13212.POREA 0 EUR
CHMPFL13212.POR FLUO013212-KIT FLUO013212-25G
2-Brom-4-fluorbenzonitril
2-Brom-4-fluorbenzonitril
Formel: C₇H₃BrFN
Molecular Weight: 200.01 g/mol
MDL: MFCD00672924
CAS-Nummer: 36282-26-5
EINECS: 252-949-7
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR