Allylphenylsulfon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(Allylsulfonyl)benzol
M-Clarity™ Qualitätsstufe = MQ200
06061-10ML
06061-50ML
SIAL06061-10MLEA
122
EUR
SIAL06061-10ML
SIAL06061-50ML
Allylphenylsulfon, Sigma-Aldrich®
Allylphenylsulfon
Formel:
C₉H₁₀O₂S Molecular Weight: 182,24 g/mol |
MDL:
MFCD00014739 CAS-Nummer: 16212-05-8 EINECS: 240-338-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...