3-Amino-1,2,4-triazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2,4-Triazin-3-ylamin, 1,2,4-Triazin-3-amin

M-Clarity™ Qualitätsstufe = MQ100

100625-10G 100625-50G
SIAL100625-10GEA 122 EUR
SIAL100625-10G SIAL100625-50G
3-Amino-1,2,4-triazin, Sigma-Aldrich®
3-Amino-1,2,4-triazine
Formel: C₃H₄N₄
Molecular Weight: 96,09 g/mol
MDL: MFCD00006459
CAS-Nummer: 1120-99-6
EINECS: 214-324-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR