Bis(4-hydroxyphenyl)sulfon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4,4'-Dihydroxydiphenylsulfon, 4-Hydroxyphenylsulfon, 4,4'-Sulfonyldiphenol
M-Clarity™ Qualitätsstufe = MQ200
103039-100G
103039-500G
SIAL103039-100GEA
50.8
EUR
SIAL103039-100G
SIAL103039-500G
Bis(4-hydroxyphenyl)sulfon, Sigma-Aldrich®
Bis(4-hydroxyphenyl)sulfon
Formel:
C₁₂H₁₀O₄S Molecular Weight: 250,28 g/mol |
MDL:
MFCD00002350 CAS-Nummer: 80-09-1 EINECS: 201-250-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...