Dichlormethylvinylsilan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Vinylmethyldichlorsilan
M-Clarity™ Qualitätsstufe = MQ100
104914-100G
104914-500G
SIAL104914-100GEA
107
EUR
SIAL104914-100G
SIAL104914-500G
Dichlormethylvinylsilan, Sigma-Aldrich®
Dichlormethylvinylsilan
Formel:
C₃H₆Cl₂Si Molecular Weight: 141.07 g/mol |
MDL:
MFCD00000492 CAS-Nummer: 124-70-9 EINECS: 204-710-3 UN: 2985 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...