2-Hydroxyethylmethacrylat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Methylpropensäure(2-hydroxyethyl)ester, Methacrylsäure(2-hydroxyethyl)ester

M-Clarity™ Qualitätsstufe = MQ200

128635-1KG 128635-500G
SIAL128635-1KGEA 61.4 EUR
SIAL128635-1KG SIAL128635-500G
2-Hydroxyethylmethacrylat, Sigma-Aldrich®
2-Hydroxyethylmethacrylat
Formel: C₆H₁₀O₃
Molecular Weight: 130.14 g/mol
MDL: MFCD00002863
CAS-Nummer: 868-77-9
EINECS: 212-782-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR