2-Amino-3-benzyloxypyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Benzyloxypyridin-2-amin, 3-Benzyloxy-2-pyridylamin

M-Clarity™ Qualitätsstufe = MQ100

144487-25G 144487-5G
SIAL144487-25GEA 192 EUR
SIAL144487-25G SIAL144487-5G
2-Amino-3-benzyloxypyridin, Sigma-Aldrich®
2-Amino-3-benzyloxypyridin
Formel: C₁₂H₁₂N₂O
Molecular Weight: 200,24 g/mol
MDL: MFCD00006316
CAS-Nummer: 24016-03-3
EINECS: 245-983-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR