Diisopropanolamin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
1,1'-Iminodipropan-2-ol, Bis(2-hydroxypropyl)amin
14960-1KG
14960-250G
SIAL14960-1KGEA
124
EUR
SIAL14960-1KG
SIAL14960-250G
Diisopropanolamin, Sigma-Aldrich®
Diisopropanolamin
Formel:
C₆H₁₅NO₂ Molecular Weight: 133,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004531 CAS-Nummer: 110-97-4 EINECS: 203-820-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...