2-Brom-4-fluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

CFF838.1 005470-5G 005470-25G
CHMPCFF838.1EA 522 EUR
CHMPCFF838.1 FLUO005470-5G FLUO005470-25G
2-Brom-4-fluorbenzoesäure
2-Bromo-4-fluorobenzoic acid
Formel: C₇H₄BrFO₂
Molecular Weight: 219,01 g/mol
Siedepunkt: 289 °C (1013 hPa)
Schmelzpunkt: 173…175 °C
MDL: MFCD00055370
CAS-Nummer: 1006-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR