2-Bromphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Bromphenylhydrazin Hydrochlorid, 1-(2-Bromphenyl)hydrazin Hydrochlorid, 2-Bromphenylhydrazinhydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

153621-10G 153621-50G
SIAL153621-10GEA 143 EUR
SIAL153621-10G SIAL153621-50G
2-Bromphenylhydraziniumchlorid, Sigma-Aldrich®
2-Bromphenylhydraziniumchlorid
Formel: C₆H₈BrClN₂
Molecular Weight: 223,5 g/mol
MDL: MFCD00012926
CAS-Nummer: 50709-33-6
EINECS: 256-728-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR