p-Toluolsulfonylisocyanat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Toluolsulfonylisocyanat, Tosylisocyanat

M-Clarity™ Qualitätsstufe = MQ200

189278-100G 189278-25G 189278-500G
SIAL189278-100GEA 53.7 EUR
SIAL189278-100G SIAL189278-25G SIAL189278-500G
p-Toluolsulfonylisocyanat, Sigma-Aldrich®
p-Toluolsulfonylisocyanat
Formel: C₈H₇NO₃S
Molecular Weight: 197,21 g/mol
MDL: MFCD00002030
CAS-Nummer: 4083-64-1
EINECS: 223-810-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR