Kaliumosmat(VI) Dihydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Kaliumosmium(VI)oxid Dihydrat, Kaliumosmat Dihydrat, Dikaliumosmat Dihydrat
M-Clarity™ Qualitätsstufe = MQ200
209104-100MG
209104-2.5G
209104-500MG
SIAL209104-100MGEA
52.6
EUR
SIAL209104-100MG
SIAL209104-2.5G
SIAL209104-500MG
Kaliumosmat(VI) Dihydrat, Sigma-Aldrich®
Kaliumosmat(VI) Dihydrat
Formel:
K₂O₄Os·2H₂O Molecular Weight: 368.45 g/mol |
MDL:
MFCD00149919 CAS-Nummer: 10022-66-9 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...