1,4-Bis(trimethylsilyl)-1,3-butadiin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,4-Bis(trimethylsilyl)buta-1,3-diyne, 1,4-Bis(trimethylsilyl)butadiyne
263567-25G
263567-5G
SIAL263567-25GEA
642
EUR
SIAL263567-25G
SIAL263567-5G
1,4-Bis(trimethylsilyl)-1,3-butadiin, Sigma-Aldrich®
1,4-Bis(trimethylsilyl)-1,3-butadiin
Formel:
C₁₀H₁₈Si₂ Molecular Weight: 194,42 g/mol |
MDL:
MFCD00009839 CAS-Nummer: 4526-07-2 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...