2,5-Dimethoxybenzolthiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Dimethoxybenzol-1-thiol, 2,5-Dimethoxybenzolthiol, 1,4-Dimethoxy-2-mercaptobenzol, 2,5-Dimethoxyphenylmercaptan

M-Clarity™ Qualitätsstufe = MQ100

275425-1G 275425-5G
SIAL275425-1GEA 77.4 EUR
SIAL275425-1G SIAL275425-5G
2,5-Dimethoxybenzolthiol, Sigma-Aldrich®
2,5-Dimethoxythiophenol
Formel: C₈H₁₀O₂S
Molecular Weight: 170,23 g/mol
MDL: MFCD00052738
CAS-Nummer: 1483-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR