Titanyl(IV)acetylacetonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Titan(IV)oxidacetylacetonat, Oxobis(2,4-pentandionato-O,O')titan(IV), Titan(IV)oxyacetylacetonat, Titan(IV)oxidbis(2,4-pentandionat)
M-Clarity™ Qualitätsstufe = MQ100
330833-10G
330833-50G
SIAL330833-10GEA
128
EUR
SIAL330833-10G
SIAL330833-50G
Titanyl(IV)acetylacetonat, Sigma-Aldrich®
Titanyl(IV)acetylacetonat
Formel:
C₁₀H₁₄O₅Ti Molecular Weight: 262.1 g/mol |
MDL:
MFCD00013505 CAS-Nummer: 14024-64-7 EINECS: 237-861-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...