Chlordimethylvinylsilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dimethylvinylchlorsilan, Vinyldimethylchlorsilan

MQ100

395439-100ML 395439-25ML
SIAL395439-100MLEA 193 EUR
SIAL395439-100ML SIAL395439-25ML
Chlordimethylvinylsilan, Sigma-Aldrich®
Chlordimethylvinylsilan
Formel: C₄H₉ClSi
Molecular Weight: 120.65 g/mol
MDL: MFCD00018090
CAS-Nummer: 1719-58-0
EINECS: 217-007-1
UN: 2985
ADR: 3 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR