Methylheptadecanoat, Supelco®

Lieferant: Merck

Synonyme: Heptadecansäuremethylester, Methylmargarat, Margarinsäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

51633-1G 51633-5G
51633-1GEA 96.7 EUR
51633-1G 51633-5G
Methylheptadecanoat, Supelco®
Methylheptadecanoat
Formel: C₁₈H₃₆O₂
Molecular Weight: 284,48 g/mol
MDL: MFCD00009001
CAS-Nummer: 1731-92-6
EINECS: 217-055-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba35cdd3aa2b51eafe9d3ac1f95e273968b41034&vid=fdee138dd473c4d9e98716e27d795d19079e7586&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR