5-Brom-2-ethoxypyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

646326-1G 646326-5G
SIAL646326-1GEA 36.2 EUR
SIAL646326-1G SIAL646326-5G
5-Brom-2-ethoxypyridin, Sigma-Aldrich®
5-Brom-2-ethoxypyridin
Formel: C₇H₈BrNO
Molecular Weight: 202,05 g/mol
MDL: MFCD00234311
CAS-Nummer: 55849-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR