Methyl-2-brom-5-methoxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Brom-5-methoxybenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

659290-25G 659290-5G
SIAL659290-25GEA 125 EUR
SIAL659290-25G SIAL659290-5G
Methyl-2-brom-5-methoxybenzoat, Sigma-Aldrich®
Methyl-2-brom-5-methoxybenzoat
Formel: C₉H₉BrO₃
Molecular Weight: 245.07 g/mol
MDL: MFCD00051594
CAS-Nummer: 35450-36-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR