(2-Bromphenyl)methylsulfon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-2-(methylsulfonyl)benzol

M-Clarity™ Qualitätsstufe = MQ100

669334-10G 669334-1G
SIAL669334-10GEA 427 EUR
SIAL669334-10G SIAL669334-1G
(2-Bromphenyl)methylsulfon, Sigma-Aldrich®
(2-Bromphenyl)methylsulfon
Formel: C₇H₇BrO₂S
Molecular Weight: 235,1 g/mol
MDL: MFCD07368924
CAS-Nummer: 33951-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR