(2-Fluorphenyl)methylsulfon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Fluor-2-(methylsulfonyl)benzol

M-Clarity™ Qualitätsstufe = MQ100

669385-10G 669385-1G
SIAL669385-10GEA 50.3 EUR
SIAL669385-10G SIAL669385-1G
(2-Fluorphenyl)methylsulfon, Sigma-Aldrich®
(2-Fluorphenyl)methylsulfon
Formel: C₇H₇FO₂S
Molecular Weight: 174,2 g/mol
MDL: MFCD03094199
CAS-Nummer: 654-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR