Methyl-3-amino-4-methoxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Amino-4-methoxybenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

683736-25G 683736-5G
SIAL683736-25GEA 118 EUR
SIAL683736-25G SIAL683736-5G
Methyl-3-amino-4-methoxybenzoat, Sigma-Aldrich®
Methyl-3-amino-4-methoxybenzoat
Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
MDL: MFCD00017196
CAS-Nummer: 24812-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR