Strontiumacetylacetonat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Strontium-2,4-pentandionat Hydrat, Bis(2,4-pentandionato-O,O')strontium Hydrat, Strontiumbis(2,4-pentandionato-O,O') Hydrat

M-Clarity™ Qualitätsstufe = MQ100

704512-25G 704512-5G
SIAL704512-25GEA 228 EUR
SIAL704512-25G SIAL704512-5G
Strontiumacetylacetonat Hydrat, Sigma-Aldrich®
Strontiumacetylacetonat Hydrat
Formel: C₁₀H₁₄O₄Sr·H₂O
Molecular Weight: 303,85 g/mol
MDL: MFCD00013502
CAS-Nummer: 12193-47-4
EINECS: 235-366-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR