3-Hydroxy-5-nitrobenzoesäure, Sigma-Aldrich®

Lieferant: Merck

714402-1G 714402-5G
SIAL714402-1GEA 61.6 EUR
SIAL714402-1G SIAL714402-5G
3-Hydroxy-5-nitrobenzoesäure, Sigma-Aldrich®
3-Hydroxy-5-nitrobenzoic acid
Formel: C₇H₅NO₅
Molecular Weight: 183,12 g/mol
MDL: MFCD01098212
CAS-Nummer: 78238-14-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR