2-Amino-3-brom-5-fluorpyridin, Sigma-Aldrich®

Lieferant: Merck

722383-1G 722383-5G
SIAL722383-1GEA 29.1 EUR
SIAL722383-1G SIAL722383-5G
2-Amino-3-brom-5-fluorpyridin, Sigma-Aldrich®
2-Amino-3-brom-5-fluorpyridin
Formel: C₅H₄BrFN₂
Molecular Weight: 191 g/mol
MDL: MFCD03092924
CAS-Nummer: 869557-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR