4-Brom-2-fluorbenzoesäure

Lieferant: FLUOROCHEM

CFF844.1 008669-5G 008669-25G
CHMPCFF844.1EA 522 EUR
CHMPCFF844.1 FLUO008669-5G FLUO008669-25G
4-Brom-2-fluorbenzoesäure
4-Bromo-2-fluorobenzoic acid
Formel: C₇H₄BrFO₂
Molecular Weight: 219,01 g/mol
MDL: MFCD00143263
CAS-Nummer: 112704-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR