Vanadyl(IV)acetylacetonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Oxobis(2,4-pentandionato-O,O')vanadium(IV), Vanadin(IV)oxyacetylacetonat, Vanadium(IV)oxidbis(2,4-pentandionat), Vanadin(IV)oxidacetylacetonat
M-Clarity™ Qualitätsstufe = MQ200
94735-250G
94735-50G
SIAL94735-250GEA
177
EUR
SIAL94735-250G
SIAL94735-50G
Vanadyl(IV)acetylacetonat, Sigma-Aldrich®
Vanadyl(IV)acetylacetonat
Formel:
C₁₀H₁₄O₅V Molecular Weight: 265,16 g/mol |
MDL:
MFCD00000032 CAS-Nummer: 3153-26-2 EINECS: 221-590-8 UN: 3285 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...