Colecalciferol (Vitamin D3), Sigma-Aldrich®
Lieferant: Merck
Synonyme: Calciol, (3β,5Z,7E)-9,10-Secocholesta- 5,7,10(19)-trien-3-ol, Vitamin D3, 3β,5-cis,7-trans-3-Hydroxy-9,10-secocholesta- 5,7,10(19)-trien, Colecalciferol, 3-[2-[7a-Methyl-1-(6-methylheptan-2-yl)- 2,3,3a,5,6,7-hexahydro-1H-inden-4-yliden] ethyliden]-4-methyliden-cyclohexan-1-ol
M-Clarity™ Qualitätsstufe = MQ200
C1357-1G C1357-250MG
SIALC1357-1GEA 190 EUR
SIALC1357-1G SIALC1357-250MG
Colecalciferol (Vitamin D3), Sigma-Aldrich®
Colecalciferol (Vitamin D3)
Formel: C₂₇H₄₄O
Molecular Weight: 384.65 g/mol
MDL: MFCD00078131
CAS-Nummer: 67-97-0
EINECS: 200-673-2
UN: 2811
ADR: 6.1 II