4-Chlorphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Chlorphenylhydrazinhydrochlorid, 4-Chlorphenylhydrazin Hydrochlorid, 1-(4-Chlorphenyl)hydrazin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

C65807-25G C65807-5G
SIALC65807-25GEA 89.1 EUR
SIALC65807-25G SIALC65807-5G
4-Chlorphenylhydraziniumchlorid, Sigma-Aldrich®
4-Chlorphenylhydraziniumchlorid
Formel: C₆H₈Cl₂N₂
Molecular Weight: 179.05 g/mol
MDL: MFCD00012943
CAS-Nummer: 1073-70-7
EINECS: 214-030-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR