3-Ethoxy-1-propanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Trimethylenglycolmonoethylether
M-Clarity™ Qualitätsstufe = MQ100
E7401-5G
E7401-25G
SIALE7401-5GEA
120
EUR
SIALE7401-5G
SIALE7401-25G
3-Ethoxy-1-propanol, Sigma-Aldrich®
3-Ethoxy-1-propanol
Formel:
C₅H₁₂O₂ Molecular Weight: 104,15 g/mol |
MDL:
MFCD00002948 CAS-Nummer: 111-35-3 EINECS: 203-861-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...